Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

UniSampler®, Sampler with Suction Lance, Bürkle

Supplier: Bürkle

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

5314-1000
10217-060EA 571.56 USD InStock
10217-060
UniSampler®, Sampler with Suction Lance, Bürkle
Samplers Fluid Samplers
UniSampler with suction lance makes sure liquid only comes into contact with the suction hose and the original sample bottle.

  • Suction tube PP, suction hose PVC
  • Universal bottle adapter made of PVC, for bottle necks dia. 24 mm, 32 mm, 39 mm and 50 mm nominal dimension

The collection lance consists of an inner tube and an outer tube. The suction position can be varied steplessly with the outer tube within a height of 0 to 48 cm.The total length of the telescopic suction tube is 100 cm.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty