Would you like to log in again?
Zone Samplers, Bürkle
Supplier: Bürkle
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5316-0055 5316-1055 5316-2055 5317-2085 5316-3150 5319-1250 5316-0150 5317-1085 5319-1150 5319-2085 5317-1058 5316-1085 5317-2055 5319-2150 5317-1150 5316-3055 5319-1085 5319-2250 5316-0085 5316-1150 5316-2150 5316-2085 5317-2150
10217-116EA 2020.56 USD InStock
10217-116 10217-106 10217-114 10217-104 10217-118 10217-108 10217-090 10217-082 10217-072 10217-080 10217-070 10217-086 10217-098 10217-076 10217-084 10217-074 10217-096 10217-068 10217-112 10217-102 10217-110 10217-100 10217-078
Zone Samplers, Bürkle
Samplers Solid Samplers
For sampling bulk goods, cross-sectional samples can be taken from different sample depths. Suitable for use with a range of sample substrates from fine powder to coarse grain.
  • Easy to clean, removable tip
  • Colour coding (Multi and Uno samplers)
  • Ultra-pure materials: Stainless steel V4A (1.4404)/AISI 316 L, PTFE or aluminium
  • 3 lengths from 550 to 1500 mm, Jumbo up to 2500 mm
  • Samplers have smallest possible gap between inner and outer tubes
Multi sampler has three or five separate chambers for taking samples at multiple depths in one action. Uno sampler has one chamber for taking a sample at one specified depth. All layer sampler has no chambers, three or five inlets and this allows one sample to be taken at multiple depths. Jumbo sampler has no chambers and three, five or seven inlets; with a 50 mm tube diameter this is ideal for larger grain sizes and quantities.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty