Would you like to log in again?
Samplers for Liquids, TeleScoop, Bürkle
Supplier: Bürkle
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5354-4100 5355-0600 5355-0120 5354-2000 5355-0300 5355-0250 5355-0280 5354-0050 5354-0100 5354-5050 5355-0450 5354-5100 5354-5200 5354-0200 5354-3000
10217-202EA 165.21 USD InStock
10217-202 10217-192 10217-182 10217-190 10217-180 10217-196 10217-174 10217-186 10217-194 10217-172 10217-184 10217-200 10217-198 10217-176 10217-188
Samplers for Liquids, TeleScoop, Bürkle
Samplers Fluid Samplers
Samplers are made from polypropylene and aluminium
  • Rigid cold rolled profile rod
  • End stop prevents rods slipping out
  • Springs made of stainless steel
For sampling from pools, tanks, manholes or surface water. Samples can be taken from as deep as 6 meters using the appropriate adjustable aluminium telescopic rod. The rods can be equipped with different attachments using the practical snap-on joint (adjustable angle beaker, pendulum beaker, bottle holder, stainless steel beaker and catch net).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty