Would you like to log in again?
Drum Pumps, Bürkle
Supplier: Bürkle
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5600-1502 5600-1501 5600-0652 5600-0801 5600-1251 5600-0651 5600-0802 5600-1002 5600-1001 5600-0502 5600-1252 5600-0501
10217-216EA 347.68 USD InStock
10217-216 10217-356 10217-214 10217-208 10217-206 10217-218 10217-354 10217-222 10217-212 10217-220 10217-352 10217-210
Drum Pumps, Bürkle
Pumps Drum/Transfer Pumps
Drum Pumps made of PP available with rigid discharge tube or flexible PVC discharge hose with stopcock
It enables fast, simple, decanting of liquids from drums, canisters and tanks. The low level base non return valve ensures good removal of the liquid remaining in the barrel. The pump is suitable for decanting acids, alkalis and aggressive cleaning agents as it has a very good chemical resistance. The delivery rate is up to 12 L/min. The flexible PVC discharge hose is 1.25 M long.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty