Would you like to log in again?
Drum Pumps for Combustible Liquids, Bürkle
Supplier: Bürkle
5601-0501 5601-0400 5601-0801 5601-0500 5601-0401 5601-0800
10217-226EA 890.48 USD InStock
10217-226 10217-224 10217-228 10217-230 10217-234 10217-232
Drum Pumps for Combustible Liquids, Bürkle
Pumps Drum/Transfer Pumps
Drum Pumps for Combustible Liquids made of Stainless steel V2A (1.4301) / AISI 304 available with rigid discharge tube or flexible 1.2 m PTFE/stainless steel discharge hose with ball valve made of stainless steel
Note: The pumps are not suitable for use with acetone.
The PTFE seals ensure long-term maintenance-free usage. The pump can conduct electricity and when used in conjunction with the anti-static kit (sold separately) fulfils the requirements for handling flammable liquids. Delivery rate depending on length of pump approximately 220 to 560 ml/stroke. The stepless adjustment of pump immersion depth is to ensure good residual extraction.
Accessories information: When decanting flammable liquids without the machines and containers involved in the process being sufficiently grounded, electrostatic build-up can occur, which for many substances can result in fire or an explosion. Burkle’s anti-static set (sold separately) ensures easy and reliable grounding. The anti-static set consists of three color-coded connecting copper cables with clamps in a handy plastic case.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty