Would you like to log in again?
Hand-Operated Solvent Pump, Bürkle
Supplier: Bürkle
5603-2000 5603-1000
10217-238EA 1057.04 USD InStock
10217-238 10217-242
Hand-Operated Solvent Pump, Bürkle
Pumps Drum/Transfer Pumps
Hand-operated solvent pump for contamination-free removal of organic solvents and flavouring agents
  • Medium-contacting parts made of stainless steel (1.4301) and PTFE
  • Rigid discharge tube with stopcock made of stainless steel/PTFE
  • Suitable for containers with R 2” thread
For containers up to 60 L. The pump is gas-tight and completely sealable with two ball valves.
Accessories information: The accessory Thread Adapter (sold separately) may be used to change a 38 mm thread of a canister to a 2" BSP thread. Suitable for US tin foil canisters with an opening of 38 mm. Adapter must be purchased separately.
When decanting flammable liquids without the machines and containers involved in the process being sufficiently grounded, electrostatic build-up can occur, which for many substances can result in fire or an explosion.
Burkle’s anti-static set (sold separately) ensures easy and reliable grounding.
The anti-static set consists of three color-coded connecting copper cables with clamps in a handy plastic case.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty