Would you like to log in again?
Wall Mounted Draining Racks, Bürkle
Supplier: Bürkle
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9640-2040 9640-2050 9640-1050 9640-2060 9640-1040 9640-1060
10217-314EA 466.74 USD InStock
10217-314 10217-312 10217-318 10217-308 10217-316 10217-310
Wall Mounted Draining Racks, Bürkle
Racks Draining Racks
  • Will not shatter or break
  • Resistant to most chemicals
  • Removable drainage channel with connection for a draining tube hose (for drainage of dripping water into laboratory basin)
This flexible PVC system has stable drainage pegs, available in three lengths (60, 100 and 150 mm) with Ø 6, 10 and 12 mm, that can be repositioned at any time. The draining racks can be adapted to any apparatus which is to be dried.
Ordering information: Rack supplied complete with removable dust- and scratch-proof protective film, and rods, screws and dowels for wall mounting.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty