Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Tubing Clamps, PVDF, Bürkle
Supplier: Bürkle
8619-0204 8619-0104 8619-0106 8619-0155 8619-0102 8619-0205 8619-0206 8619-0152 8619-0105 8619-0202 8619-0156 8619-0154
10217-390EA 11.58 USD InStock
10217-390 10217-394 10217-384 10217-274 10217-392 10217-272 10217-382 10217-398 10217-388 10217-396 10217-276 10217-386
Tubing Clamps, PVDF, Bürkle
Clamps Tubing and Sanitary Clamps
The micro adjustment screw enables the flow of liquids to be controlled delicately and smoothly or stopped completely.
  • PVDF with galvanised steel hook
  • Heat resistant up to +160 °C
  • Various sizes available for tubing from 1 - 20 mm external diameter
  • Available in four colours
The clamp can also be mounted on existing tube connections. The clamp's closure can be opened in any position, without screwing on and off.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty