Would you like to log in again?
Metal Hub Needles, Hamilton
Supplier: Hamilton
91024 91020 91025 91021
10217-424PK 91.36 USD InStock
10217-424 10217-422 10217-428 10217-426
Metal Hub Needles, Hamilton
Syringe Needles
Metal hub (nickel plated brass) needles can be used with TLL syringes and LT or TLL connectors.
  • Autoclavable
  • 304 stainless steel needles
  • Custom needle lengths and point styles are available
Packaging: Conveniently packed with 6 needles per pack.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty