Would you like to log in again?
Kel-F® Hub Needles, Hamilton
Supplier: Hamilton
90525 90528 90532 90530 90524 90516 90533 90531 90517 90514 90539 90521 90523 90534 90520 90535 90536
10217-468PK 127.52 USD InStock
10217-468 10217-488 10217-478 10217-456 30618-692 10217-490 10217-482 10217-460 10217-472 10217-480 10217-470 10217-464 10217-442 10217-476 10217-484 10217-462 10217-474
Kel-F® Hub Needles, Hamilton
Syringe Needles
Kel-F® hub (KF) needles can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors.
  • Autoclavable, gas sterilizable
  • 304 stainless steel needles
Packaging: Conveniently packed with 6 needles per pack.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty