Would you like to log in again?
Quantofix Carbonate Hardness Test Strips, CTL Scientific Supply
Supplier: CTL Scientific Supply
91323
10217-496EA 58.11 USD InStock
10217-496
Quantofix Carbonate Hardness Test Strips, CTL Scientific Supply
Environmental Test Kits
Quantofix Carbonate Hardness Test strips for the semi-quantitative determination of the carbonate hardness
Quantofix Carbonate Hardness Test strips have a gradation of scale: 0-3-6-10-15-20 mg/L light green to blue. Strong bases or other acid-consuming substances cause false results.
Carbonate hardness is that part of the total hardness resulting from the content of alkaline earth ions, which are equivalent to the carbonate and hydrogen carbonate ions present in the water. Thus the carbonate hardness is the portion of the total hardness which contributes to the formation of alkaline earth carbonates (boiler scale).
Ordering information: Packaged as a box of 100.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty