Would you like to log in again?
Quantofix Chloride Test Strips, CTL Scientific Supply
Supplier: CTL Scientific Supply
91321
10217-498EA 72.19 USD InStock
10217-498
Quantofix Chloride Test Strips, CTL Scientific Supply
Environmental Test Kits
Quantofix Chloride Test strips test for the semi-quantitative determination of chloride (Cl -) in solutions
Quantofix Chloride Test strips has a gradation of scale: 0-500-1000-1500-2000 mg/l Cl- brown to yellow. The following ions only interfere above the indicated concentrations: >1000 mg/L Al3+, Ca2+, Cd2+, Ce4+, Cu2+, K+, Mg2+, Mn2+, Na+, NH4+, Ni2+, Pb2+, Sn2+, Zn2+, BO3 -, CO3 2- CrO 4 2-, NO2 -, NO 3 -, PO43-, SO32-, SO42-, acetate, citrate, oxalate, tartrate, EDTA > 500 mg/L Fe2+, Fe3+ > 300 mg/L l-, >100 mg/L Ag+ .
In the pH range 2 to 12 the reaction is independent of the pH value of the solution to be analyzed. Strongly acidic solutions have to be adjusted to pH 5 - 7 with sodium hydroxide solution, alkaline solutions are adjusted to the same pH range with nitric acid. Chloride concentrations greater than 3000 mg/L can be determined after defined dilution of the test sample with distilled water.
Ordering information: Packaged as a box of 100.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty