Would you like to log in again?
Quantofix Arsenic Test Kits, CTL Scientific Supply
Supplier: CTL Scientific Supply
91334 91345 91332
10217-502EA 156.18 USD InStock
10217-502 10217-506 10217-504
Quantofix Arsenic Test Kits, CTL Scientific Supply
Environmental Test Kits
Quantofix Arsenic test kits and reagents for the semi-quantitative determination of arsenic in solutions
Quantofix Arsenic 10 has a gradation of scale: 0-0.01-0.025-0.05-0.1-0.5 ppm.
The addition of zinc and a solid acid results in the formation of arsine from any arsenic compounds in the sample solution. Arsine reacts in the gaeous phase with the mercury bromide in the reaction zone of the test stick to form mixed arsenic/mercury halides, which, depending on the concentration, are colored yellow to brown.
Ordering information: Packaged as a box of 100, strips and reagent.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty