Would you like to log in again?
Quantofix Cyanide Test Kit, CTL Scientific Supply
Supplier: CTL Scientific Supply
91318
10217-514EA 118.97 USD InStock
10217-514
Quantofix Cyanide Test Kit, CTL Scientific Supply
Environmental Test Kits
Quantofix Cyanide test strips and reagents for the semi-quantitative determination of cyanide in solutions.
Quantofix Cyanide has a gradation of scale: 0-1-3-10-30 mg/L.
Ordering information: Packaged as a box of 100, strips and reagent.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty