Would you like to log in again?
Quantofix Formaldehyde Test Kit, CTL Scientific Supply
Supplier: CTL Scientific Supply
91328
10217-516EA 99.6 USD InStock
10217-516
Quantofix Formaldehyde Test Kit, CTL Scientific Supply
Environmental Test Kits
Quantofix Formaldehyde test strips and reagents for the semi-quantitative determination of formaldehyde in solutions
Quantofix Formaldehyde test kit has a gradation of scale: 0-10-20-40-60-100-200 mg/L HCHO beige to blue-violet.
Other aldehydes such as e.g. acetaldehyde or glutardialdehyde only react at higher concentrations, resulting in a discoloration which deviates from the color scale. Acetone does not react. Strong oxidizing and reducing agents cause low results.
Ordering information: Packaged as a box of 100, strips and reagent.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty