Would you like to log in again?
Quantofix Phosphate Test Kit, CTL Scientific Supply
Supplier: CTL Scientific Supply
91320
10217-552EA 88.16 USD InStock
10217-552
Quantofix Phosphate Test Kit, CTL Scientific Supply
Environmental Test Kits
Quantofix Phosphate test strips and reagent for the semi-quantitative determination of ortho-phosphte (PO 4 3-) in solutions
Quantofix Phosphate test strips has a gradation of scale: 0-3-10-25-50-100 mg/L PO43- white to blue-green.
Only Ortho-phosphate is determined. Other phosphates, such as poly-, pyro-, and metaphosphates, have to be decomposed for the determination of total phosphates. Silica (SiO 2) contents above 10 mg/L react in the same way giving a blue coloration. Larger amounts of sulphide ions (S 2) cause a brown color of the test field, smaller concentrations cause low results. The following ions do not interfere: 1000 mg/L Ag +, Al 3+, Cd 2+, Co 2+, Cr 3+, Mg 2+, Mn 2+, NH 4+, Ni 2+, Zn 2+, Cl-, F -, NO 3-, SO 4 2-, citrate, oxalate, tartrate, <500 mg/L Ca 2+, < 250 mg/L Pb 2+, <50 mg/L Cu 2+ <25 mg/L Fe 3+, 5 mg/L Fe 2+, <2 mg/L NO 2 -.
Ordering information: Packaged as a box of 100, strips and reagent.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty