Would you like to log in again?
Quantofix Sulfate Test Strips, CTL Scientific Supply
Supplier: CTL Scientific Supply
91329
10217-558EA 60.82 USD InStock
10217-558
Quantofix Sulfate Test Strips, CTL Scientific Supply
Environmental Test Kits
Semi-quantitative test strips for the determination of Sulfate. Ideal for rapid and easy analysis of sulfate directly on-site.
  • No maintenance or calibration is required.
Ordering information: Packaged as a box of 100.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty