Would you like to log in again?
Quantofix QUAT Test Strips, CTL Scientific Supply
Supplier: CTL Scientific Supply
91337
10217-572EA 55.99 USD InStock
10217-572
Quantofix QUAT Test Strips, CTL Scientific Supply
Environmental Test Kits
Quantofix QUAT test strips for the rapid and reliable determination of quaternary ammonium compounds (QUAT) in solutions.
Quantofix Quat has a gradation of scale: 0-10-25-50-100-250-500-1000 mg/L.
Ordering information: Packaged as a box of 100.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty