Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Recombinant Lipoamide Dehydrogenase (from E. coli)

Supplier: Novus Biologicals
NBP1-41232
102170-048EA 465.66 USD InStock
102170-048
Human Recombinant Lipoamide Dehydrogenase (from E. coli)
Enzymes
The Recombinant Human Dihydrolipoamide Dehydrogenase/DLD Protein from Novus Biologicals is derived from E. coli. The Recombinant Human Dihydrolipoamide Dehydrogenase/DLD Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

  • Entrez Gene ID: 1738

Tested Applications: ELISA, SDS-Page. Tested Reactivity: Human.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty