Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Nalgene® Syringe Filters, Surfactant-Free Cellulose Acetate and Cellulose Acetate, 25mm, Thermo Scientific
Supplier: Thermo Scientific
723-9920 723-2520 724-2020 723-2545 724-2045 723-2580 724-2080 723-9945
10218-476CS 669.64 USD InStock
10218-476 10218-478 10218-480 10218-482 10218-470 10218-484 10218-472 10218-474
Nalgene® Syringe Filters, Surfactant-Free Cellulose Acetate and Cellulose Acetate, 25mm, Thermo Scientific
Filters Syringe Filters
Filter sample volumes of 10 to 100 ml with these 25 mm Ø syringe filters.
  • Sterile filters are certified sterile, non-cytotoxic and non-pyrogenic
  • SFCA syringe filters have a blue housing ring
CA membranes have some protein binding and extractables, making them more suitable for clarification of buffers, aqueous solutions and reagents. CA syringe filters have a red housing ring. All housings are polypropylene with female Luer-Lok®inlet and male Luer slip outlet.
SFCA membranes are low protein binding and have lower extractables than standard CA so SFCA is a good choice for cell culture fluids and cellular solutions.
Packaging: Sterile filters are individually blister-packed. Non-sterile filters are bulk packed in zip closure bags.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty