Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Nalgene® Syringe Filters, Polyethersulfone, 25mm, Thermo Scientific
Supplier: Thermo Scientific
725-2520 725-2545
10218-488CS 275.19 USD InStock
10218-488 10218-486
Nalgene® Syringe Filters, Polyethersulfone, 25mm, Thermo Scientific
Filters Syringe Filters
Filter aqueous solutions and media with Thermo Scientific™ Nalgene™ 25 mm Ø Syringe Filters with PES (polyethersulfone) membrane.
  • Accommodate sample volumes from 10 to 100 ml
These filters are certified sterile, non-cytotoxic and non-pyrogenic. They have a polyethersulfone (PES) membrane with polypropylene housing. Individually blister-packed. The inlet is a female Luer-Lok® and the outlet is a male Luer-Slip. Green housing ring.
PES is the best choice for filtering cell culture media and fluids because of its low protein binding and extractables.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty