Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Nalgene® Syringe Filters, Nylon, 25mm, Thermo Scientific

Supplier: Thermo Scientific
727-2020 727-2045 726-2520 726-2545
10218-490CS 333.41 USD InStock
10218-490 10218-492 10218-494 10218-496
Nalgene® Syringe Filters, Nylon, 25mm, Thermo Scientific
Filters Syringe Filters
These 25 mm Ø Nylon syringe filters are ideal for filtering samples and mobile phases used in GC and HPLC.

  • Sterile filters are certified sterile, non-cytotoxic and non-pyrogenic
  • Accommodate sample volumes from 10 to 100 ml

Nylon membrane is naturally hydrophilic, has low extractables and some protein binding. The nylon membrane and polypropylene housing are chemically-resistant to a wide range of solvents. Pink housing ring.

They are also useful for filtering alcohols, cryoprotectants such as DMSO, aqueous/biological solutions and weak solvents.

Packaging: Sterile filters are individually blister-packed. Non sterile filters are bulk packed in zip closure bags.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty