Would you like to log in again?
TOC Cleaning Validation Kits, Texwipe®
Supplier: Texwipe
TX3342 TX3343 TX3340
TWTX3342EA 739.73 USD InStock
TWTX3342 TWTX3340 10218-590
TOC Cleaning Validation Kits, Texwipe®
Swabs Swabs for Cleaning
Texwipe’s Cleaning Validation Kits were designed to simplify sampling as part of a cleaning validation protocol. The swabs and vials are further cleaned using a proprietary process to make these kits excellent for use in Total Organic Carbon (TOC) analysis.
  • Swab head is cleanroom laundered, double-layered polyester knit
  • Break-away handle allows the swab head to be placed into a sampling vial with minimal handling
Kits contain vials, swabs, and labels to sample multiple places in the production area. Prewashed TOC vials have been manufactured, analyzed, and certified to virtually elimnate all background TOC. Swab has a large head that provides firm support for sampling broad areas and flat surfaces. Can be used with Sievers Model 800/900, Anatel ANATOC™, Teledyne Tekmar, Ol Analytical Model 1010 and Shimadzu TOC-V Series™ TOC Analyzers.
Packaging: Kits are packaged in cleanroom-compatible polypropylene boxes for efficient transport of samples from production to laboratory with minimal chance of contamination.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty