Would you like to log in again?
BLAUBRAND® Graduated Pipettes, Class AS, USP, Certified, BrandTech
Supplier: BrandTech
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BLAUBRAND®
27506 27511 27509 27513
10218-686PK 138.81 USD InStock
10218-686 10218-688 10218-690 10218-684
BLAUBRAND® Graduated Pipettes, Class AS, USP, Certified, BrandTech
Pipettes Graduated Pipettes
Volumetrically stable even when heated up to 250 °C via special annealing process.
  • Certified Class A
  • Calibrated to deliver (TD, Ex.)
  • Constructed according to DIN EN ISO standards with error limits that meet the specifications of USP/ASTM
  • Flexible screen stenciled graduations for maximal accuracy at intermediate volumes
  • Durable, high-contrast blue enamel markings
Each individual piece is marked on a computer controlled, automated production line to ensure maximum precision.
Certifications: Class AS per DIN EN ISO 835. Meets standards of USP Section 31. AQL of 0.4 per DIN ISO 3951.
Delivery information: USP batch certificates are supplied.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty