Would you like to log in again?
BRAND® BLAUBRAND® Volumetric Flasks, Class AS, USP, Certified, with Glass Stopper, BrandTech® Scientific
Supplier: BrandTech
Brand®
36981 36983 36979 36975 36968 36982 36984 36980 36977 36978 36973
10218-696PK 80.52 USD InStock
10218-696 10218-710 10218-698 10218-712 10218-700 10218-702 10218-704 10218-706 10218-692 10218-708 10218-694
BRAND® BLAUBRAND® Volumetric Flasks, Class AS, USP, Certified, with Glass Stopper, BrandTech® Scientific
Flasks Volumetric Flasks
BRAND® BLAUBRAND® volumetric flasks are certified Class AS and constructed to DIN EN ISO standards of Boro 3.3 glass and meet the error limits of USP/ASTM.
  • Individually adjusted
  • Durable, highly legible BLAUBRAND® ink (blue line) for easy adjustment and maximum precision
High quality inks are baked on during an annealing process that relieves thermal stresses. This provides volume stability even after heating up to 250 °F. Statistical process control ensures compliance with predefined specifications of at least 99.6%. Calibrated to contain (TC, In). Made in BRAND's Wertheim Germany facility.
Certifications: DIN EN ISO 1042.
Ordering information: Includes one USP batch certificate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty