Would you like to log in again?
Biox Cryogenic Storage Boxes
Supplier: TradeWinds (TWD)
Biox
BX98-0113 BX98-0213
10218-762CS 253.32 USD InStock
10218-762 10218-760
Biox Cryogenic Storage Boxes
Boxes Cryoboxes
Cryogenic storage boxes are manufactured from polycarbonate for liquid nitrogen and deep freezer use.
  • Clear removable lid
  • Numbered grip
  • Drain holes
  • 1 and 2" options available
Always in a 10×10 configuration. Openings in the bottom make them scan ready when used with 2D barcoded vials.
Ordering information: Storage boxes are packaged 12 per case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty