Would you like to log in again?
Seven2Go™ S4 Portable Dissolved Oxygen Meter, METTLER TOLEDO®
Supplier: Mettler-Toledo
Seven2Go™
30207960 30207959 30207958
10218-830EA 1135 USD InStock
10218-830 10218-832 10218-834
Seven2Go™ S4 Portable Dissolved Oxygen Meter, METTLER TOLEDO®
Electrochemical Meters Oxygen Meters
These robust meters with polarographic or optical sensor provide durability and one-handed operation, making them ideally suited for mobile applications in the laboratory, at-line, or outdoors.
  • New intuitive menu guarantees ease of use
  • T-Pad with smart button arrangement enables easy and fast navigation
  • Instrument automatically recognises InLab ISM sensors and stores sensor ID, serial number, and last calibration data
  • Automatic or manual temperature and pressure compensation
Thanks to their IP 67 protection and drop test resistance, the devices are able to withstand harsh and demanding environments.
Ordering information: The S4 meter only is supplied with electrode clip, operating instructions, quick guide, test certificate, declaration of conformity and batteries.
S4 Standard Kit: As S4 meter, but also with InLab 605-ISM and bottles for calibration.
S4 Field Kit: As S4 Standard Kit, but including uGo™ carrying case and zero oxygen tablets.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty