Would you like to log in again?
Seven2Go™ S7 Pro Portable Conductivity Meter, METTLER TOLEDO®
Supplier: Mettler-Toledo
Seven2Go™ S7 Pro
30207962 30207873 30207961 30207963
10218-840EA 2265 USD InStock
10218-840 10218-842 10218-836 10218-838
Seven2Go™ S7 Pro Portable Conductivity Meter, METTLER TOLEDO®
Electrochemical Meters Conductivity Meters
These robust meters provide durability and one-handed operation, making them ideally suited for mobile applications in the laboratory, at-line, or outdoors.
  • New intuitive menu guarantees ease of use
  • T-Pad with smart button arrangement enables easy and fast navigation
  • Instrument automatically recognises InLab ISM sensors and stores sensor ID, serial number, and last calibration data
  • Linear temperature compensation
  • Screen's contrast is enhanced by direct sunlight: Excellent readability even under bright and sunny conditions
  • StatusLight shows the user instantly if the meter is ready or not
  • Data storage of 2000 values (GLP conform)
  • Linear or non linear temperature compensation
  • Acoustic endpoint signal
Thanks to their IP67 protection and drop test resistance, the devices are able to withstand harsh and demanding environments.
Ordering information: The S7 Pro meter only includes LabX direct pH, USB cable, electrode clip, base accessory, operating instructions, quick guide, test certificate, declaration of conformity and batteries.
S7 Standard Kit: As S7-Meter, but also with InLab 738-ISM, bottles for calibration and sachets of conductivity standards 1413 µS/cm and 1288 mS/cm.
S7 Field Kit: As S7-Standard kit, but also with uGo™ carrying case.
S7 USP/EP Kit: As S7 Field Kit, but with InLab 742-ISM instead of InLab 738-ISM.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty