Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water, distilled

Supplier: GROWCELLS
Synonyms: Dihydrogen oxide

SDW-1000 SDW-010Q SDW-0125 SDW-0500 SDW-55GL SDW-010L SDW-2000 SDW-020Q
75788-556EA 325.72 USD InStock
75788-556 75788-554 10220-376 10218-874 10218-900 10218-902 75788-552 75788-550
Water, distilled
Water
Used in the preparation of cell culture media and laboratory reagents.

  • Cell culture quality
  • Molecular biological grade
  • Prep laboratory reagents

Used in the preparation of cell culture media and laboratory reagents. Cell culture quality and molecular biological grade.

Ordering information: Water and buffers in IV bags are not injectable. They are not a medical device and are not to be used for intravenous use. They are for research only.


Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty