Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Proteasome 20S LMP2 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-52090
102182-652EA 564.74 USD InStock
102182-652
Anti-Proteasome 20S LMP2 Goat Polyclonal Antibody
Antibodies
Goat Polyclonal Proteasome 20S LMP2 Antibody. Tested Applications: Western Blot, Peptide ELISA. Tested Reactivity: Human.

Type: Primary
Antigen: Proteasome 20S LMP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty