Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water, Cell Culture Grade

Supplier: GROWCELLS
Synonyms: Dihydrogen oxide

CCPW-0125 CCPW-2000 CCPW-0500 CCPW-020Q CCPW-010L CCPW-1000
75788-538CS 693.87 USD InStock
75788-538 10218-876 75788-536 10218-880 10220-414 75788-540
Water, Cell Culture Grade
Water
Distilled cell culture grade water.

  • Prepared by distillation
  • Steam sterilization in final packaging
  • 0.1 micron sterile filtration

Distilled cell culture grade water, <0.005EU/mL. Prepared by distillation, 0.1 micron sterile filtration and steam sterilization in final packaging.


Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty