Would you like to log in again?
Pro10 Field pH/ORP Handheld Meter, YSI
Supplier: YSI
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6050010
10221-846EA 852.1 USD InStock
10221-846
Pro10 Field pH/ORP Handheld Meter, YSI
Electrochemical Meters pH and Multi-Parameter Meters
Rugged and reliable, the YSI Pro10 provides everything you need in a handheld instrument for the measurement of either pH or ORP along with temperature.
User-replaceable sensors and cables along with convenient calibration routine feature makes it extremely user friendly. Mil-spec (military spec) connectors, IP67 waterproof and rubber over molded case, and 1-meter drop tests ensure the instrument remains in your hands to provide years of sampling even in the harshest field conditions. The feature set of the Pro10 results in a low total cost of ownership over the life of the product and proves that it is smarter, tougher, and more versatile.
The Pro10 simultaneously measures pH/ORP, and temperature with the following features. User-replaceable cables and sensors provide versatility, reduce down time and reduce overall cost of ownership. Available with 1- 4- 10- 20- and 30-m cable lengths standard (cable management kit included on all cables 4-m and longer). Smart calibration; the instrument remembers the previous calibration values and simply walks you through the routine. Stores 50 data sets; no need to write down data. Large graphic, backlit display and glow in the dark keypad. Usable in any lighting condition. The display is also visible when wearing polarized sunglasses.Tough. IP67, impact-resistant, waterproof case. Waterproof without the battery cover as well. Rubber over molded case provides extra durability. Military spec connectors are unsurpassed for ruggedness. Multiple languages include English, Spanish, French, and Portuguese. 3-year instrument; 2-year cable; 1-year sensor warranty
Certifications: RoHS, CE, WEEE, IP-67, 1-meter drop test. Waterproof to IP67 (even with the battery cover off).
Ordering information: When ordering the Pro10 you must order the instrument, cable, and pH or ORP sensor separately, allowing you to choose the system that best fits your application. Temperature is part of the cable you order and included in the cable purchase. Contact your VWR representative for more information. Three-year instrument warranty; 2-year cable; 1-year pH and ORP sensors.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty