Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Proteasome 20S LMP2 Mouse Monoclonal Antibody [clone: 1D1]

Supplier: Novus Biologicals
NBP2-01817
102230-370EA 663.82 USD InStock
102230-370
Anti-Proteasome 20S LMP2 Mouse Monoclonal Antibody [clone: 1D1]
Antibodies
Mouse Monoclonal Proteasome 20S LMP2 Antibody (1D1). Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry. Tested Reactivity: Human.

Type: Primary
Antigen: Proteasome 20S LMP2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty