Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Staphylococcus Enterotoxin B Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-10722R-HRP
10245-606EA 811.1 USD InStock
10245-606
Anti-Staphylococcus Enterotoxin B Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Staphylococcal enterotoxins represent a group of proteins, which are secreted by Staphylococcus aureus and cause staphylococcal food poisoning syndrome. The illness is characterised by high fever, hypotension, diarrhea, shock, and in some cases death. Their molecular masses range between 27 and 30 kDa. At present, seven enterotoxins are known, namely A, B, C1, C2, C3, D and E. Their amino acid sequences have been determined and it was shown that all are single chain polypeptides containing one disulfide bond formed by two half cystines located in the middle of the polypeptide chain, which form the so called cysteine loop. SEB is an extremely potent activator of T cells, stimulating the production and secretion of various cytokines which mediate many of the toxic effects of SEB.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: Staphylococcus Enterotoxin B
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty