Would you like to log in again?
Artificial Eccrine Perspiration, Pickeringlab
Supplier: Pickering Laboratories
1700-0024
102465-390EA 126.85 USD InStock
102465-390
Artificial Eccrine Perspiration, Pickeringlab
Educational Materials Biology Educational Materials Human Biology Learning Activities
Artificial eccrine perspiration is a ready-to-use solution and is the closest mimic to true human eccrine sweat.
  • Universally standardize
  • Used in research and product testing
  • Formula that can satisfy all test challenges
  • Reliable, reproducible and consistent solutions
It consists of 19 amino acids, the seven most abundant minerals, and the four most abundant metabolites at a pH of 4.5. All concentrations closely match experimentally determined values for adult human eccrine sweat.
The stabilized solution is preserved with a fungicide and bactericide for two years of shelf life at room temperature.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty