Would you like to log in again?
StabilCoat® BSA ELISA Stabilizer/Blocker, Surmodics
Supplier: Surmodics
StabilCoat®
SC01-0050 SC01-1000 SC01-2000
102466-300EA 436.15 USD InStock
102466-300 102466-298 102466-296
StabilCoat® BSA ELISA Stabilizer/Blocker, Surmodics
Buffers Electrophoresis Buffers Blocking Buffers
StabilCoat® Immunoassay Stabilizer, and the 5X concentrate, effectively preserves the conformation and activity of dried proteins while simultaneously blocking the surface to reduce non-specific binding. The product can be used to stabilize antibodies, antigens or enzymes on an assortment of immunoassay components including polystyrene plates, tubes, glass, membranes, and filter paper.
  • 3 year shelf life
  • Increased stability
  • Reduces non-specific binding
StabilCoat® Immunoassay Stabilizer increases signal-to-noise ratios of assays through effective blocking of the plate while maximizing the binding capacity of the antigen/antibody epitopes.
Certifications: ISO 13485:2016 & 9001:2015 Certified.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty