Would you like to log in again?
StabilGuard® Immunoassay Stabilizer, Surmodics IVD, Inc.
Supplier: Surmodics
StabilGuard®
SG01-1000 SG01-0050
102466-310EA 43.08 USD InStock
102466-310 102466-312
StabilGuard® Immunoassay Stabilizer, Surmodics IVD, Inc.
Buffers Electrophoresis Buffers Blocking Buffers
StabilGuard® Immunoassay Stabilizer (BSA-Free) effectively preserves the conformation and activity of dried proteins while simultaneously blocking the surface to reduce non-specific binding. It can be used to stabilize antibodies, antigens or enzymes on an assortment of immunoassay components including polystyrene plates, tubes, glass, membranes, and filter paper.
  • BSA/Protein Free
  • Increased Stability
  • Reduced non-specific binding
  • 3 year stability
The key benefit of the protein free formulation is the elimination of interference and cross reactivity within immunoassay applications.
StabilGuard® (BSA free) Immunoassay Stabilizers increase signal-to-noise ratios of assays through effective blocking of the plate while maximizing the binding capacity of the antigen/antibody epitopes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty