Would you like to log in again?
Human Recombinant Efnb1 Cc54
Supplier: Bon Opus Biosciences
CC54-50UG CC54-10UG CC54-500UG CC54-1MG
102497-504EA 2487.69 USD InStock
102497-504 102497-502 102497-508 102497-506
Human Recombinant Efnb1 Cc54
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human Ephrin-B1 is produced by our mammalian expression system and the target gene encoding Leu28-Gly232 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped via FedEx.
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty