Would you like to log in again?
Human Fam172A Ci26
Supplier: Bon Opus Biosciences
CI26-10UG CI26-1MG CI26-50UG CI26-500UG
102499-812EA 283.23 USD InStock
102499-812 102499-810 102499-808 102499-806
Human Fam172A Ci26
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant Human Protein FAM172A is produced by our Mammalian expression system and the target gene encoding Gln19-Lys416 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx.
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty