Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
NEBNEXT® Small RNA Library Prep Set for Illumina (Multiplex Compatible), New England Biolabs
Supplier: New England Biolabs
NEBNext®
E7330S E7330S E7330L E7330L
102500-114EA 1285 USD InStock
102500-114 CA102500-114 102500-112 CA102500-112
NEBNEXT® Small RNA Library Prep Set for Illumina (Multiplex Compatible), New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The novel NEBNext® Small RNA workflow has been optimized to minimize adaptor-dimers while producing high-yield, high-diversity libraries
The NEBNext® Small RNA Library Prep Set for Illumina® (Multiplex Compatible) enables use with your own barcode system. The kit contains adaptors but does not include multiplex primers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty