Would you like to log in again?
E. faecalis O-Glycosidase and Neuraminidase Bundle
Supplier: New England Biolabs
E0540S E0540S
102500-120EA 221 USD InStock
CA102500-120 102500-120
E. faecalis O-Glycosidase and Neuraminidase Bundle
Enzymes
1 set of this bundle includes 2,000,000 units of O-Glycosidase and 2,000 units of Neuraminidase
O-Glycosidase, also known as Endo-α-N-Acetylgalactosaminidase, catalyzes the removal of Core 1 and Core 3 O-linked disaccharides from glycoproteins. Neuraminidase is the common name for Acetyl-neuraminyl hydrolase (Sialidase). This Neuraminidase catalyzes the hydrolysis of α2-3, α2-6, and α2-8 linked N-acetyl-neuraminic acid residues from glycoproteins and oligosaccharides.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty