Would you like to log in again?
Phusion Hot Start Flex 2X Master Mix, New England Biolabs
Supplier: New England Biolabs
Phusion®
M0536S M0536L M0536S M0536L
102500-126EA 238 USD InStock
102500-126 CA102500-126 102500-124 CA102500-124
Phusion Hot Start Flex 2X Master Mix, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Phusion Hot Start Flex 2X Master Mix offers robust high fidelity performance and room temperature reaction setup in a convenient master mix format
The addition of an aptamer-based inhibitor allows room temperature reaction setup. The convenient master mix formulation is supplied at a 2X concentration and contains dNTPs and Mg++, requiring only the addition of primers and DNA template for robust amplification. Reactions can also be optimized using the provided DMSO solution.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty