Would you like to log in again?
Phusion Hot Start Flex DNA Polymerase, New England Biolabs
Supplier: New England Biolabs
Phusion®
M0535S M0535L M0535S M0535L
102500-128EA 639 USD InStock
CA102500-128 102500-128 102500-130 CA102500-130
Phusion Hot Start Flex DNA Polymerase, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Phusion Hot Start Flex DNA Polymerase offers robust, high fidelity performance and room temperature reaction setup
The addition of an aptamer-based inhibitor allows room temperature reaction setup. Phusion Hot Start Flex DNA Polymerase is supplied with standard 5X Phusion HF Buffer, as well as 5X Phusion GC Buffer, which can be used for complex or GC-rich templates.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty