Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Q5® High-Fidelity DNA Polymerase, New England BioLabs
Supplier: New England Biolabs
Q5®
M0491L M0491S M0493S M0492S M0494X E0555S E0555S M0494X M0491L M0494S E0555L E0555L M0494L M0493L M0491S M0494S M0493L M0492L M0492L M0494L M0493S M0492S
102500-138EA 128 USD InStock
102500-138 102500-146 102500-136 102855-122 102500-140 102855-120 102500-144 102500-134 102500-142 102500-132 CA76307-458 CA102500-138 76307-458 CA102500-142 CA102500-132 CA102500-140 CA102500-146 CA102855-122 CA102500-136 CA102855-120 CA102500-144 CA102500-134
Q5® High-Fidelity DNA Polymerase, New England BioLabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
The Q5® High-Fidelity 2X Master Mix offers robust, high-fidelity performance in a convenient master mix format.
  • For use in high-specificity PCR, high-fidelity PCR, cloning, long or difficult amplification and high-throughput PCR.
The Q5 High- Fidelity 2X Master Mix features a high-fidelity, thermostable DNA polymerase with 3´→ 5´ exonuclease activity, fused to a processivity-enhancing Sso7d domain to support robust DNA amplification. With an error rate > 100-fold lower than that of Taq DNA Polymerase and 12-fold lower than that of Pyrococcus furiosus (Pfu) DNA Polymerase, Q5 High-Fidelity DNA Polymerase is ideal for cloning and can be used for long or difficult amplicons. The convenient master mix formulation is supplied at a 2X concentration. The mix contains dNTPs, Mg++ and a proprietary broad-use buffer requiring only the addition of primers and DNA template for robust amplification regardless of GC content. When used at the recommended 1X final concentration, the Q5 High-Fidelity Master Mix contains 2 mM MgCl2. Q5 High-Fidelity DNA Polymerase is unlike typical, lower fidelity PCR enzymes.
Ordering information: The Q5 High-Fidelity DNA Polymerase and the Q5 Hot Start High-Fidelity DNA Polymerase are supplied with Q5 High GC Enhancer and Q5 Reaction Buffer.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty