Would you like to log in again?
SpeI-HF®, Restriction Enzymes, New England Biolabs
Supplier: New England Biolabs
R3133M R3133M
102500-166EA 323 USD InStock
102500-166 CA102500-166
SpeI-HF®, Restriction Enzymes, New England Biolabs
Nucleic Acid Reagents Restriction Enzymes
An E. coli strain that carries the cloned and modified SpeI gene from Sphaerotilus species (ATCC 13923). High Fidelity (HF®) Restriction Enzymes have 100% activity in CutSmart® Buffer; single-buffer simplicity means more straightforward and streamlined sample processing. HF enzymes also exhibit dramatically reduced star activity. HF enzymes are all Time-Saver™ qualified and can therefore cut substrate DNA in 5-15 with the flexibility to digest overnight without degradation to DNA. Engineered with performance in mind, HF restriction enzymes are fully active under a broader range of conditions, minimizing off-target products, while offering flexibility in experimental design.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty