Would you like to log in again?
Human Recombinant TGF beta 1 (from CHO Cells)
Supplier: AdipoGen Life Sciences
CHI-HF-210TGFBM-C100
102511-372EA 609.23 USD InStock
102511-372
Human Recombinant TGF beta 1 (from CHO Cells)
Proteins and Peptides
TGF-beta1 (Transforming growth factor beta 1) is a polypeptide member of the transforming growth factor beta superfamily of cytokines. It is a secreted protein that performs many cellular functions, including the control of cell growth, cell proliferation, cell differentiation and apoptosis. In humans, TGF-beta1 is encoded by the TGFB1 gene. TGF-beta1 was first identified in human platelets as a protein with a molecular mass of 25 kaD with a potential role in wound healing. It was later characterized as a large protein precursor (containing 390 aa) that was proteolytically processed to produce a mature peptide of 112 aa. TGF-beta1 plays an important role in controlling the immune system, and shows different activities on different types of cell, or cells at different developmental stages. Most immune cells (or leukocytes) secrete TGF-beta1.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty