Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FTO Mouse Monoclonal Antibody [clone: AG103]

Supplier: AdipoGen Life Sciences
AG-20A-0092-C050 AG-20A-0092-C100
102514-252EA 387.69 USD InStock
102514-252 102514-250
Anti-FTO Mouse Monoclonal Antibody [clone: AG103]
Antibodies
FTO (Fat mass-and obesity-associated gene) is the responsible gene for mouse 'fused toes' mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower HDL-cholesterol.

Type:
Antigen: FTO
Clonality: Monoclonal
Clone: AG103
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty