Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Leptin Rabbit Polyclonal Antibody

Supplier: AdipoGen Life Sciences
AG-25A-0009-C100
102514-398EA 353.08 USD InStock
102514-398
Anti-Leptin Rabbit Polyclonal Antibody
Antibodies
Leptin is involved in the signaling pathway that regulates the size of the body fat depot. An increase in the level of leptin may act directly or indirectly on the CNS to inhibit food intake and/or regulate energy expenditure as part of a homeostatic mechanism to maintain constancy of the adipose mass. Defects in the leptin gene may be a cause of autosomal recessive obesity.

Type:
Antigen: Leptin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty