Would you like to log in again?
BAFF, Soluble (Human) Matched Pair Detection Set
Supplier: AdipoGen Life Sciences
AG-46B-0001-KI01
102515-636EA 900 USD InStock
102515-636
BAFF, Soluble (Human) Matched Pair Detection Set
Assays ELISAs
BAFF (B cell-activating factor; BLyS; B lymphocyte stimulator; TNFSF13B; TALL-1; CD257), belonging to the TNF family, is a master regulator of peripheral B cell survival, and together with IL-6, promotes Ig class-switching and plasma cell differentiation
BAFF co-stimulates activated T cells. Increased levels of soluble BAFF have been detected in the serum of patients with various autoimmune diseases, such as Sjögren's syndrome, rheumatoid arthritis, multiple sclerosis and systemic lupus erythematosus (SLE). Furthermore, BAFF is found in inflammatory sites in which there is lymphoid neogenesis. BAFF levels are elevated in patients with multiple myeloma and B cell chronic lymphoid leukemia (B-CCL).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty