Would you like to log in again?
17α-Hydroxyprogesterone, 17a-OHP, Human ELISA, BioVendor
Supplier: BioVendor
RCD021R
102552-042EA 580.8 USD InStock
102552-042
17α-Hydroxyprogesterone, 17a-OHP, Human ELISA, BioVendor
Assays ELISAs
Measurment of 17αOHP is therefore valuable in the initial diagnosis of CAH.
  • Calibration Range: 0.15-20 ng/mL
  • Limit of Detection: 0.11 ng/mL
  • Serum: 50 uL/well
The steroid 17α−OH Progesterone is produced by the adrenal cortex and gonads. The 17α−OH Progesterone has little progestational activity, but it is of intense clinical interest because it is the immediate precursor to 11-desoxycortisol (Cpd-S). Cpd-S is produced by 21-hydroxylation of 17 OHP, measurement of 17aOHP is a useful indirect indicator of 21-hydroxlase activity. In congenital 21– hydroxylase deficiency, the most common variety of congenital adrenal hyperplasia (CAH), 17αOHP is secreted in abundant excess. Measurment of 17αOHP is therefore valuable in the initial diagnosis of CAH.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty